Learn More
Eurofins Discoverx Recombinant Human IL-21, 10 µg. (92-1287)
Supplier:  Eurofins Discoverx 921287
Encompass
Interleukin 21 (IL-21) is a cytokine protein and mediator of the immune response. Induces differentiation, proliferation, and activity of macrophages, NK, B, and cytotoxic T cells in both innate and adaptive immune responses. Synergizes with IL-15 and IL-18, and is implicated in multiple immune-mediated diseases, including lupus, psoriasis, and chronic inflammatory diseases. Agonist to IL21R. Provided as 10 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9804