Learn More
Eurofins Discoverx Recombinant Human IL-24, 20 µg. (92-1288)
Supplier:  Eurofins Discoverx 921288
Encompass
Interleukin 24 (IL-24) is a pro-apoptotic cytokine protein of the IL10 family that is a mediator of the immune response. It is recognized as a tumor-suppressing protein due to its role in control of cell survival and proliferation through activation of the STAT1 and STAT3 transcription factors. Important roles in wound healing, psoriasis, and cancer tissues. Agonist for IL20RA and IL22RA1. Provided as 200 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9805