Learn More
Eurofins Discoverx Recombinant Human IL-26, 25 µg. (92-1274)
Supplier:  Eurofins Discoverx 921274
Encompass
Interleukin 26 (IL-26) is a cytokine protein and mediator of the immune response that is overexpressed in herpesvirus samimiri-transformed T cells, and thought to contribute to the transformed phenotype of T cells after herpesvirus infection. IL-26 is an agonist for IL20RA, has roles in mucosal immunity, and may be a pro-inflammatory factor. Il-26 is inhibited by heparin, and may be implicated in inflammatory bowel disease. It is similar in sequence to IL-10. Provided as 25 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9791