Learn More
Eurofins Discoverx Recombinant Human IL-29, 20 µg. (92-1257)
Supplier:  Eurofins Discoverx 921257
Encompass
Interferon lambda 1 (IFN-λ1, IL-29) is a cytokine protein and mediator of the immune response. Highly upregulated in viral infections, and stimulates MHC class I antigen expression. Agonist of the heterodimeric class II cytokine receptors IL10RA and IL28RB. Provided as 200 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9777