Learn More
Eurofins Discoverx Recombinant Human IL-4, 20 µg. (92-1254)
Supplier:  Eurofins Discoverx 921254
Encompass
Interleukin 4 (IL-4) is an anti-inflammatory cytokine protein and mediator of the immune response. Agonist of the interleukin 4 receptor. Activates B-cells during an inflammatory response and induces differentiation of helper T cells to Th2 cells. It is a co-stimulator of DNA-synthesis, and enhances secretion and expression of IgE and IgG1. Provided as 200 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9775