Learn More
Eurofins Discoverx Recombinant Human IL-9, 10 µg. (92-1271)
Supplier:  Eurofins Discoverx 921271
Encompass
Interleukin 9 (IL-9) is a cytokine protein and mediator of the immune response. Produced by CD4+ T helper cells, it stimulates cell proliferation, prevents apoptosis, and regulates hematopoietic cells. IL-9 acts as an agonist of the interleukin-9 receptor (IL9R) and activates the STAT proteins. It is implicated in asthma and the pathogenesis of bronchial hyperresponsiveness. Provided as 10 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9789