Learn More
Eurofins Discoverx Recombinant Human Leptin, 200 µg. (92-1327)
Supplier:  Eurofins Discoverx 921327
Encompass
Leptin is an adipose-derived cytokine hormone encoded by the ob (obese) gene and an agonist for LEPR. Leptin is involved in regulating fat stores, blood glucose levels, and food consumption as well as regulating energy balance, suppressing the appetite, and increasing thermogenesis. Provided as 200μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9839