Learn More
Eurofins Discoverx Recombinant Human PDGF-BB, 2 µg. (92-1110)
Supplier:  Eurofins Discoverx 921110
Encompass
Platelet-derived growth factor BB (PDGF-BB) is a homodimeric protein that is released upon platelet activation that induces proliferation, migration, and survival of endothelial, epithelial, and smooth muscle cells, as well as fibroblasts. A potent mitogen for mesenchymal cells, PDGF-BB is able to bind PDGFRa and PDGFRb, and plays an important part in healing, regeneration, angiogenesis and formation of smooth muscle during embryonic development. May facilitate tumor metastasis and vascularization. Provided as 2 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9645