Learn More
Eurofins Discoverx Recombinant Human TNF alpha, 10 µg. (92-1097)
Supplier:  Eurofins Discoverx 921097
Encompass
Tumor necrosis factor alpha (TNFα) is an endogenous pyrogen cytokine that plays a key part in regulating immune cells, immune response to infection, and systemic inflammation. Also regulates cell proliferation, differentiation, apoptosis, lipid metabolism, and coagulation. Is implicated in multiple autoimmune disorders due to its role in inflammation, including inflammatory bowel disease, rheumatoid arthritis, ankylosing spondylitis, and psoriasis. Agonist of TNF receptor 1 and 2. Provided as 10 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9634