Learn More
Eurofins Discoverx Recombinant Human TSLP, 10 µg. (92-1276)
Catalog No. 501999793
Encompass
Thymic stromal lymphopoietin (TSLP) is a cytokine protein and agonist for IL7R. It is responsible for inducing the release of T cell-attracting chemokines from monocytes and enhancing maturation of myeloid dendritic cells, such as Langerhans cells. TSLP induces allergic inflammation by directly activating mast cells. Provided as 10 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9793 Supplier Eurofins Discoverx Supplier No. 921276
Thymic stromal lymphopoietin (TSLP) is a cytokine protein and agonist for IL7R. It is responsible for inducing the release of T cell-attracting chemokines from monocytes and enhancing maturation of myeloid dendritic cells, such as Langerhans cells. TSLP induces allergic inflammation by directly activating mast cells. Provided as 10 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.