Learn More
ATCC Pneumococcal polysaccharide powder Type 2 (d2) 2mg
Catalog No. 502381515
Encompass
Purified pneumococcal polysaccharide type 2 is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal polysaccharide type 2 can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-1515 Supplier ATCC Supplier No. 500X
Purified pneumococcal polysaccharide type 2 is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal polysaccharide type 2 can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.