Learn More
Medchemexpress LLC RI-AG03 acetate | 98.2% | 2477.93 | 1 MG
SDP
Catalog No. 5000366078
Encompass_Preferred
RI-AG03 acetate is the acetate of RI-AG03 (HY-P10861), an orally active and BBB-penetrable Tau aggregation peptide inhibitor. It inhibits Tau aggregation and improves associated neurodegeneration and behavioral phenotypes in both in vivo and in vitro models, making it useful in the study of tauopathies such as Alzheimer's disease.
  • Inhibits tau aggregation
  • Improves associated neurodegeneration and behavioral phenotypes
  • Orally active and BBB-penetrable
  • Suitable for in vivo and in vitro studies
  • Useful in the study of tauopathies
Catalog No. 50-003-66078 Supplier Medchemexpress LLC Supplier No. 1MGHYP10861A
RI-AG03 acetate is the acetate of RI-AG03 (HY-P10861), an orally active and BBB-penetrable Tau aggregation peptide inhibitor. It inhibits Tau aggregation and improves associated neurodegeneration and behavioral phenotypes in both in vivo and in vitro models, making it useful in the study of tauopathies such as Alzheimer's disease.
  • Inhibits tau aggregation
  • Improves associated neurodegeneration and behavioral phenotypes
  • Orally active and BBB-penetrable
  • Suitable for in vivo and in vitro studies
  • Useful in the study of tauopathies