Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC RI-AG03 acetate | 98.2% | 2477.93 | 10 MG
SDP

Catalog No. 5000365093
Encompass_Preferred

RI-AG03 acetate is an orally active and BBB-penetrable Tau aggregation peptide inhibitor. It inhibits Tau aggregation and improves associated neurodegeneration and behavioral phenotypes in both in vivo and in vitro models. It can be used in the study of tauopathies such as Alzheimer's disease.

  • Inhibits Tau aggregation
  • Improves associated neurodegeneration and behavioral phenotypes
  • Orally active and BBB-penetrable
  • Suitable for the study of tauopathies such as Alzheimer's disease
  • Molecular weight: 2477.93 (free base)
  • Appearance: Solid
  • Color: White to off-white
  • Formula: C104H189N49O22·xC2H4O2

Catalog No. 50-003-65093 Supplier Medchemexpress LLC Supplier No. 10MGHYP10861A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RI-AG03 acetate is an orally active and BBB-penetrable Tau aggregation peptide inhibitor. It inhibits Tau aggregation and improves associated neurodegeneration and behavioral phenotypes in both in vivo and in vitro models. It can be used in the study of tauopathies such as Alzheimer's disease.

  • Inhibits Tau aggregation
  • Improves associated neurodegeneration and behavioral phenotypes
  • Orally active and BBB-penetrable
  • Suitable for the study of tauopathies such as Alzheimer's disease
  • Molecular weight: 2477.93 (free base)
  • Appearance: Solid
  • Color: White to off-white
  • Formula: C104H189N49O22·xC2H4O2

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.