Learn More
Medchemexpress LLC RI-AG03 acetate | 98.2% | 2477.93 | 5 MG
SDP
Catalog No. 5000393444
Encompass_Preferred
RI-AG03 acetate is the acetate of RI-AG03, an orally active and BBB-penetrable Tau aggregation peptide inhibitor. It inhibits Tau aggregation and improves associated neurodegeneration and behavioral phenotypes in both in vivo and in vitro models. This product can be used in the study of tauopathies such as Alzheimer's disease.
  • Orally active
  • BBB-penetrable Tau aggregation peptide inhibitor
  • Inhibits Tau aggregation
  • Improves associated neurodegeneration and behavioral phenotypes in both in vivo and in vitro models
  • Can be used in the study of tauopathies such as Alzheimer's disease
Catalog No. 50-003-93444 Supplier Medchemexpress LLC Supplier No. 5MGHYP10861A
RI-AG03 acetate is the acetate of RI-AG03, an orally active and BBB-penetrable Tau aggregation peptide inhibitor. It inhibits Tau aggregation and improves associated neurodegeneration and behavioral phenotypes in both in vivo and in vitro models. This product can be used in the study of tauopathies such as Alzheimer's disease.
  • Orally active
  • BBB-penetrable Tau aggregation peptide inhibitor
  • Inhibits Tau aggregation
  • Improves associated neurodegeneration and behavioral phenotypes in both in vivo and in vitro models
  • Can be used in the study of tauopathies such as Alzheimer's disease