Learn More
Waters Corp Waters Corporation Refractive Index Qualification Test Solution Kit, RI Detector Testing, Multi-Concentration Caffeine
Catalog No. NC1830612 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Refractive Index Qualification Test Solution is designed for use with refractive index (RI) detectors to test their sensitivity, reproducibility, and linearity. It is intended for use by a certified engineer or a user as part of a performance maintenance or qualification activity.
  • Intended for use with refractive index (RI) detectors.
  • Used to test the sensitivity, reproducibility, and linearity of the refractive index detector.
  • Designed for use by a certified engineer or user for performance maintenance or qualification.
  • Protocols for use are available through the purchase of qualification services or self-directed qualification training and license.
  • Contains caffeine at concentrations of 0.1 mg/mL, 0.2 mg/mL, 0.5 mg/mL, 1 mg/mL, and 10 mg/mL.
  • Manufactured in state-of-the-art facilities adhering to cGMP and ISO 9001 conditions.
Catalog No. NC1830612 Supplier Waters Corp Supplier No. 186007208
The Refractive Index Qualification Test Solution is designed for use with refractive index (RI) detectors to test their sensitivity, reproducibility, and linearity. It is intended for use by a certified engineer or a user as part of a performance maintenance or qualification activity.
  • Intended for use with refractive index (RI) detectors.
  • Used to test the sensitivity, reproducibility, and linearity of the refractive index detector.
  • Designed for use by a certified engineer or user for performance maintenance or qualification.
  • Protocols for use are available through the purchase of qualification services or self-directed qualification training and license.
  • Contains caffeine at concentrations of 0.1 mg/mL, 0.2 mg/mL, 0.5 mg/mL, 1 mg/mL, and 10 mg/mL.
  • Manufactured in state-of-the-art facilities adhering to cGMP and ISO 9001 conditions.