Learn More
Medchemexpress LLC RI(dl)-2 TFA | 98.0% | 1 MG
SDP
Catalog No. 5000341318
Encompass_Preferred
RI(dl)-2 TFA is a potent and selective RAD51-mediated D-loop formation inhibitor.
  • Potent and selective RAD51-mediated D-loop formation inhibitor with an IC50 of 11.1 μM.
  • Does not influence RAD51 binding to ssDNA.
  • Inhibits homologous recombination (HR) activity in human cells (IC50 of 3.0 μM).
Catalog No. 50-003-41318 Supplier Medchemexpress LLC Supplier No. HY126972A1MG
RI(dl)-2 TFA is a potent and selective RAD51-mediated D-loop formation inhibitor.
  • Potent and selective RAD51-mediated D-loop formation inhibitor with an IC50 of 11.1 μM.
  • Does not influence RAD51 binding to ssDNA.
  • Inhibits homologous recombination (HR) activity in human cells (IC50 of 3.0 μM).