Learn More
Medchemexpress LLC RI(dl)-2 TFA | 98.0% | 1 ML
SDP
Catalog No. 5000342819
Encompass_Preferred
RI(dl)-2 TFA is a potent and selective RAD51-mediated D-loop formation inhibitor with an IC50 of 11.1 μM. It inhibits homologous recombination (HR) activity in human cells with an IC50 of 3.0 μM and does not influence RAD51 binding to ssDNA.
  • Potent and selective RAD51-mediated D-loop formation inhibitor
  • IC50 of 11.1 μM for D-loop formation
  • Inhibits homologous recombination (HR) activity in human cells
  • Does not influence RAD51 binding to ssDNA
  • Stabilizes nucleoprotein filaments in a nonfunctional state
  • Shielded from pathways promoting single-strand annealing
Catalog No. 50-003-42819 Supplier Medchemexpress LLC Supplier No. HY126972A10MM/1ML
RI(dl)-2 TFA is a potent and selective RAD51-mediated D-loop formation inhibitor with an IC50 of 11.1 μM. It inhibits homologous recombination (HR) activity in human cells with an IC50 of 3.0 μM and does not influence RAD51 binding to ssDNA.
  • Potent and selective RAD51-mediated D-loop formation inhibitor
  • IC50 of 11.1 μM for D-loop formation
  • Inhibits homologous recombination (HR) activity in human cells
  • Does not influence RAD51 binding to ssDNA
  • Stabilizes nucleoprotein filaments in a nonfunctional state
  • Shielded from pathways promoting single-strand annealing