Learn More
Microsolv Technology Corporation Performance Qualification Solutions, RI. Set of Solutions for Qualification of HPLC with RI Detectors. Chemical Solutions Brand. 1 EA.
Supplier:  Microsolv Technology Corporation 49102RI
Encompass_Preferred
This set of NIST Traceable Solutions are Supplemental for use with the HSQ™ or PQ™ Kits for Performance Qualification of HPLC instruments to Qualify Refractive Index (RI) Detectors. Certificate of Analysis is included with each set. Set Includes: 6 x 30ml Linearity Solutions 1 x 30ml Diluent Blank Solution Solutions will be supplied with at least six months until the expiration date. They are designed to be used promptly and often. Expiration date is two years after the date of manufacture.
Catalog No. 50-226-9766