Learn More
TARGETMOL CHEMICALS INC RIAA-94 5MG
SDP
Catalog No. 502581421
Encompass_Preferred
Also available in 2 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. R(+)-IAA-94 (R(+)-Methylindazone) is a potent indanyloxyacetic acid blocker of epithelial chloride channels. R(+)-IAA-94 inhibits Nef-sdAb19 (single-domain antibody) interaction and binds to negative factor (Nef). purity: 99%
Catalog No. 50-258-1421 Supplier TARGETMOL CHEMICALS INC Supplier No. T127885MG
Also available in 2 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. R(+)-IAA-94 (R(+)-Methylindazone) is a potent indanyloxyacetic acid blocker of epithelial chloride channels. R(+)-IAA-94 inhibits Nef-sdAb19 (single-domain antibody) interaction and binds to negative factor (Nef). purity: 99%