Learn More
Med Vet International Rib Cutter/ Shearer, 9in, J0508, Each
Catalog No. 5000584723
Encompass_Preferred
Cuts through rib cage on small animals and younger large animals. German-made. Stainless steel.
Catalog No. 50-005-84723 Supplier Med Vet International Supplier No. JOR508
Cuts through rib cage on small animals and younger large animals. German-made. Stainless steel.