Learn More
World Precision Instrument Rib Cutting Knife, 40 mm
SDP
Catalog No. 502538375
Encompass
The Rib Cutting Knife is designed for gross anatomy dissection and post-mortem exams. It has a thick cutting edge and a sturdy, balance handle for cutting through ribs. Made of standard grade stainless steel, it is easily sanitized with any disinfectant or autoclaved between uses.
Catalog No. 50-253-8375 Supplier World Precision Instrument Supplier No. WP0079
The Rib Cutting Knife is designed for gross anatomy dissection and post-mortem exams. It has a thick cutting edge and a sturdy, balance handle for cutting through ribs. Made of standard grade stainless steel, it is easily sanitized with any disinfectant or autoclaved between uses.