Learn More
Alkali Scientific Ribavirin, 1 Gram
SDP
Catalog No. 5000659634
Encompass_Preferred
Ribavirin is a potent antiviral compound commonly used in laboratory settings for the study and treatment of viral infections. This chemical reagent, available in a 1g pack, has broad-spectrum activity against various RNA and DNA viruses. It works by inhibiting viral RNA synthesis, making it an essential tool in antiviral research and clinical studies. Ribavirin is used in the treatment of diseases like hepatitis C and respiratory syncytial virus (RSV). In addition to its antiviral properties, it is used in pharmaceutical formulations to help fight off viral infections by interfering with viral replication processes. This product is laboratory grade and ideal for scientific applications.
Catalog No. 50-006-59634 Supplier Alkali Scientific Supplier No. IBA001CP
Ribavirin is a potent antiviral compound commonly used in laboratory settings for the study and treatment of viral infections. This chemical reagent, available in a 1g pack, has broad-spectrum activity against various RNA and DNA viruses. It works by inhibiting viral RNA synthesis, making it an essential tool in antiviral research and clinical studies. Ribavirin is used in the treatment of diseases like hepatitis C and respiratory syncytial virus (RSV). In addition to its antiviral properties, it is used in pharmaceutical formulations to help fight off viral infections by interfering with viral replication processes. This product is laboratory grade and ideal for scientific applications.