Learn More
Medchemexpress LLC Ribavirin-13C5 | 1646818-35-0 | 1 MG
SDP
Catalog No. 5000409307
Encompass_Preferred
Ribavirin-13C5 is a 13C-labeled version of Ribavirin, an antiviral agent effective against a wide range of viruses including HCV, HIVl, and RSV. This compound is intended for research use only.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-09307 Supplier Medchemexpress LLC Supplier No. HYB0434S1MG
Ribavirin-13C5 is a 13C-labeled version of Ribavirin, an antiviral agent effective against a wide range of viruses including HCV, HIVl, and RSV. This compound is intended for research use only.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.