Learn More
Cayman Chemical Ribavirin-13C5, Purity: ≥95%, CAS: 1646818-35-0, 1 mg
SDP
Catalog No. 502910710
Encompass_Preferred
An internal standard for the quantification of ribavirin by GC- or LC-MS
Catalog No. 50-291-0710 Supplier Cayman Chemical Supplier No. 348821 MG
An internal standard for the quantification of ribavirin by GC- or LC-MS