Learn More
Alkali Scientific Ribavirin, 250 Milligrams
SDP
Catalog No. 5000659609
Encompass_Preferred
Ribavirin is a nucleoside analog and antiviral compound used primarily in the research of viral infections. This reagent is commonly utilized for studying its efficacy against viruses such as influenza and hepatitis C. Ribavirin functions by inhibiting viral RNA synthesis, thus reducing the ability of viruses to replicate. In laboratory settings, it is used to simulate viral infection and assess antiviral treatment methods. Ribavirin's broad-spectrum antiviral properties make it essential in studying a wide range of viral pathogens. This particular formulation comes in 250 milligrams for research use, packaged for laboratory applications in molecular biology and virology.
Catalog No. 50-006-59609 Supplier Alkali Scientific Supplier No. IBA002CP
Ribavirin is a nucleoside analog and antiviral compound used primarily in the research of viral infections. This reagent is commonly utilized for studying its efficacy against viruses such as influenza and hepatitis C. Ribavirin functions by inhibiting viral RNA synthesis, thus reducing the ability of viruses to replicate. In laboratory settings, it is used to simulate viral infection and assess antiviral treatment methods. Ribavirin's broad-spectrum antiviral properties make it essential in studying a wide range of viral pathogens. This particular formulation comes in 250 milligrams for research use, packaged for laboratory applications in molecular biology and virology.