Learn More
Medchemexpress LLC Ribavirin | 36791-04-5 | 100 MG
SDP
Catalog No. 5000408944
Encompass_Preferred
Ribavirin (ICN-1229) is an antiviral agent effective against a broad spectrum of viruses, including HCV, HIVl, and RSV. It also exhibits anti-orthopoxvirus and anti-variola activities, and has demonstrated activity against DENV.
  • Antiviral agent against a broad spectrum of viruses
  • Effective against HCV, HIVl, and RSV
  • Exhibits anti-orthopoxvirus and anti-variola activities
  • Reduces NO2 levels in LPS-stimulated microglia
  • Inhibits DENV virus infection in cultured cells
Catalog No. 50-004-08944 Supplier Medchemexpress LLC Supplier No. HYB0434100MG
Ribavirin (ICN-1229) is an antiviral agent effective against a broad spectrum of viruses, including HCV, HIVl, and RSV. It also exhibits anti-orthopoxvirus and anti-variola activities, and has demonstrated activity against DENV.
  • Antiviral agent against a broad spectrum of viruses
  • Effective against HCV, HIVl, and RSV
  • Exhibits anti-orthopoxvirus and anti-variola activities
  • Reduces NO2 levels in LPS-stimulated microglia
  • Inhibits DENV virus infection in cultured cells