Learn More
Medchemexpress LLC Ribavirin | 36791-04-5 | 99.9% | 1 ML
SDP
Catalog No. 5000407897
Encompass_Preferred
Ribavirin is an antiviral agent effective against a broad spectrum of viruses, including HCV, HIVl, and RSV. It also exhibits anti-orthopoxvirus and anti-variola activities. This product is for research use only.
  • Antiviral agent
  • Effective against broad spectrum of viruses, including HCV, HIVl, and RSV
  • Exhibits anti-orthopoxvirus activity
  • Exhibits anti-variola activity
  • Provided as a 10 mM solution in DMSO
  • Purity of 99.96%
Catalog No. 50-004-07897 Supplier Medchemexpress LLC Supplier No. HYB043410MM/1ML
Ribavirin is an antiviral agent effective against a broad spectrum of viruses, including HCV, HIVl, and RSV. It also exhibits anti-orthopoxvirus and anti-variola activities. This product is for research use only.
  • Antiviral agent
  • Effective against broad spectrum of viruses, including HCV, HIVl, and RSV
  • Exhibits anti-orthopoxvirus activity
  • Exhibits anti-variola activity
  • Provided as a 10 mM solution in DMSO
  • Purity of 99.96%