Learn More
Medchemexpress LLC Ribavirin | 36791-04-5 | 99.96% | 200 MG
SDP
Catalog No. 5000405333
Encompass_Preferred
Ribavirin (ICN-1229) is an antiviral agent effective against a wide range of viruses. It exhibits activity against Hepatitis C Virus (HCV), Human Immunodeficiency Virus type 1 (HIVl), and Respiratory Syncytial Virus (RSV). Additionally, Ribavirin demonstrates anti-orthopoxvirus and anti-variola activities.
  • Broad-spectrum antiviral agent targeting various viruses including HCV, HIVl, and RSV.
  • Effective against orthopoxviruses and variola.
Catalog No. 50-004-05333 Supplier Medchemexpress LLC Supplier No. HYB0434200MG
Ribavirin (ICN-1229) is an antiviral agent effective against a wide range of viruses. It exhibits activity against Hepatitis C Virus (HCV), Human Immunodeficiency Virus type 1 (HIVl), and Respiratory Syncytial Virus (RSV). Additionally, Ribavirin demonstrates anti-orthopoxvirus and anti-variola activities.
  • Broad-spectrum antiviral agent targeting various viruses including HCV, HIVl, and RSV.
  • Effective against orthopoxviruses and variola.