Learn More
Cayman Chemical Ribavirin 5'-monophosphate (lithium salt), Purity: ≥90%, CAS: 66983-94-6, 5 mg
SDP
Catalog No. 502986652
Encompass_Preferred
Competitively inhibits IMPDH in vitro (Ki = 270 nM); a metabolite of the prodrug ribavirin; broad-spectrum antiviral activity, particularly against influenza and HIV
Catalog No. 50-298-6652 Supplier Cayman Chemical Supplier No. 218215 MG
Competitively inhibits IMPDH in vitro (Ki = 270 nM); a metabolite of the prodrug ribavirin; broad-spectrum antiviral activity, particularly against influenza and HIV