Learn More
Abcam Ribavirin, Antiviral nucleoside, 10MG
Catalog No. 502707376
Encompass_Preferred
MW 244.21 g/mol, Purity >99%. A non interferon-inducing, broad spectrum antiviral nucleoside. Active metabolites interfere with viral replication. Shows antiviral activity against a number of viruses including Hepatitis C, influenza and polio.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7376 Supplier Abcam Supplier No. AB12066010MG
MW 244.21 g/mol, Purity >99%. A non interferon-inducing, broad spectrum antiviral nucleoside. Active metabolites interfere with viral replication. Shows antiviral activity against a number of viruses including Hepatitis C, influenza and polio.
The product is subject to the following: Abcam Restricted Use Statement