Learn More
Cole-Parmer Dynalon Ribbed low-density polyethylene matting, 50' long
Supplier:  Cole-Parmer 0680920
Encompass_Preferred
Versatile, virgin LDPE matting resists solvents. Use to line shelves and drawers or as a floor liner. Cushions benchtop and reduces glass breakage. Clear, ribbed; 0.036" thick through ribs.
Catalog No. NC0410655