Learn More
MOLDPRO INC Ribbed Watch Glass for 15 mL Digestion Tube, Clear, 1000/pkg, PP Homopolymer
Catalog No. NC2233835
Encompass_Preferred
MoldPro MP-128 ribbed watch glass is designed to fit our MP-123 15 mL digestion vessels.
This item is not returnable. View return policy
Catalog No. NC2233835 Supplier MOLDPRO INC Supplier No. MP128
MoldPro MP-128 ribbed watch glass is designed to fit our MP-123 15 mL digestion vessels.