Learn More
MOLDPRO INC Ribbed Watch Glass for 100 mL Digestion Tube, Clear, 500/pkg, PP Homopolymer
Catalog No. NC2233838
Encompass_Preferred
MoldPro MP-130 ribbed watch glass is designed to fit MP-125 100 mL digestion vessels.
This item is not returnable. View return policy
Catalog No. NC2233838 Supplier MOLDPRO INC Supplier No. MP130
MoldPro MP-130 ribbed watch glass is designed to fit MP-125 100 mL digestion vessels.