Learn More
Grainger Ribbon 4-1/8 in W 200 ft L
Catalog No. 502750438
Encompass_Preferred
The Grainger website offers a variety of thermal transfer ribbons for barcode and label printing. These ribbons are compatible with Zebra, Sato, Datamax, Intermec, and other popular thermal transfer printers. They come in wax, wax/resin, and resin formulations in various widths and lengths. The ribbons provide crisp, scannable barcodes and text when used with compatible labels. Selecting the right ribbon depends on the substrate, image durability, chemical/solvent resistance needed. Grainger offers a wide selection of high-quality ribbons to meet various application requirements.
Catalog No. 50-275-0438 Supplier Grainger Supplier No. 14A410
The Grainger website offers a variety of thermal transfer ribbons for barcode and label printing. These ribbons are compatible with Zebra, Sato, Datamax, Intermec, and other popular thermal transfer printers. They come in wax, wax/resin, and resin formulations in various widths and lengths. The ribbons provide crisp, scannable barcodes and text when used with compatible labels. Selecting the right ribbon depends on the substrate, image durability, chemical/solvent resistance needed. Grainger offers a wide selection of high-quality ribbons to meet various application requirements.