Learn More
Grainger Ribbon Orange 8-3/4 in W 200 ft L
Catalog No. 502751570
Encompass_Preferred
The Brady Label Printer Ribbon 8-13 is a thermal transfer printer ribbon that is compatible with Brady label printers. It features a width of 1.18 inches (30mm) and a length of 361 feet (110m). The ribbon uses a wax/resin ink that adheres to a variety of label materials including paper, polyester, vinyl, polyimide and more. It produces sharp barcodes, logos and text when used with a Brady label printer. The ribbon comes packaged one roll per box.
Catalog No. 50-275-1570 Supplier Grainger Supplier No. 14A582
The Brady Label Printer Ribbon 8-13 is a thermal transfer printer ribbon that is compatible with Brady label printers. It features a width of 1.18 inches (30mm) and a length of 361 feet (110m). The ribbon uses a wax/resin ink that adheres to a variety of label materials including paper, polyester, vinyl, polyimide and more. It produces sharp barcodes, logos and text when used with a Brady label printer. The ribbon comes packaged one roll per box.