Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ribitol-1-13C | 488-81-3 | MFCD00064291 | 99.5% | 153.14 g/mol | C413CH12O5 | 10 MG
SDP

Catalog No. 5000211578
Encompass_Preferred

Ribitol-1-13C is a 13C-labeled ribitol used as a tracer and internal standard in metabolic and analytical studies. It is a crystalline pentose alcohol supplied as a solid for precise quantitation in NMR, GC-MS, and LC-MS workflows.

  • Stable 13C label at the 1-position for tracer and quantitation studies.
  • Suitable for NMR, GC-MS, and LC-MS analytical applications.
  • High purity (99.5%) suitable for quantitative analysis.
  • Solid, white to off-white appearance for easy handling.
  • Molecular weight 153.14 g/mol (labeled isotopologue).
  • Store at -20°C, protect from light, under nitrogen; in solution: -80°C up to 6 months, -20°C up to 1 month.

Catalog No. 50-002-11578 Supplier Medchemexpress LLC Supplier No. HY100582S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ribitol-1-13C is a 13C-labeled ribitol used as a tracer and internal standard in metabolic and analytical studies. It is a crystalline pentose alcohol supplied as a solid for precise quantitation in NMR, GC-MS, and LC-MS workflows.

  • Stable 13C label at the 1-position for tracer and quantitation studies.
  • Suitable for NMR, GC-MS, and LC-MS analytical applications.
  • High purity (99.5%) suitable for quantitative analysis.
  • Solid, white to off-white appearance for easy handling.
  • Molecular weight 153.14 g/mol (labeled isotopologue).
  • Store at -20°C, protect from light, under nitrogen; in solution: -80°C up to 6 months, -20°C up to 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.