Learn More
Medchemexpress LLC Ribitol-2-13C | 488-81-3 | 98.0% | 153.14 | C4 13CH12O5 | 5 MG
SDP
Catalog No. 5000213742
Encompass_Preferred
Ribitol-2-13C is a stable isotope-labeled form of ribitol used as an internal standard and analytical reference in metabolomics, mass spectrometry, and isotope-tracing studies. It is supplied as a high-purity, accurately quantified material suitable for calibration, quantitation, and method development.
  • Stable-isotope labeled ribitol for accurate internal standardization.
  • High purity, 98.0%.
  • Supplied as a precisely weighed milligram-scale powder for analytical workflows.
  • Suitable for metabolomics, LC-MS/MS, and isotope-tracing experiments.
  • Includes documentation such as SDS and data sheet for safe handling.
Catalog No. 50-002-13742 Supplier Medchemexpress LLC Supplier No. HY100582S15MG
Ribitol-2-13C is a stable isotope-labeled form of ribitol used as an internal standard and analytical reference in metabolomics, mass spectrometry, and isotope-tracing studies. It is supplied as a high-purity, accurately quantified material suitable for calibration, quantitation, and method development.
  • Stable-isotope labeled ribitol for accurate internal standardization.
  • High purity, 98.0%.
  • Supplied as a precisely weighed milligram-scale powder for analytical workflows.
  • Suitable for metabolomics, LC-MS/MS, and isotope-tracing experiments.
  • Includes documentation such as SDS and data sheet for safe handling.