Learn More
Medchemexpress LLC Ribitol-2-13C | 488-81-3 | 98.0% | 153.14 g/mol | C413CH12O5 | 100 MG
SDP
Catalog No. 5000214757
Encompass_Preferred
Ribitol-2-13C is a stable isotope-labeled ribitol used as a tracer or internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. It is a crystalline pentose alcohol supplied for research applications.
  • 13C-labeled tracer for metabolic and quantitative studies.
  • Suitable as internal standard for NMR, GC-MS, and LC-MS.
  • High purity, ≥98.0%.
  • Molecular weight 153.14 g/mol.
  • Available in small laboratory pack sizes such as 100 mg.
Catalog No. 50-002-14757 Supplier Medchemexpress LLC Supplier No. HY100582S1100MG
Ribitol-2-13C is a stable isotope-labeled ribitol used as a tracer or internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. It is a crystalline pentose alcohol supplied for research applications.
  • 13C-labeled tracer for metabolic and quantitative studies.
  • Suitable as internal standard for NMR, GC-MS, and LC-MS.
  • High purity, ≥98.0%.
  • Molecular weight 153.14 g/mol.
  • Available in small laboratory pack sizes such as 100 mg.