Learn More
Medchemexpress LLC Ribitol-3-13C | 488-81-3 | 99.7% | 153.14 | C413CH12O5 | 5 MG
SDP
Catalog No. 5000213882
Encompass_Preferred
Ribitol-3-13C is a 13C-labeled ribitol provided as a stable isotope reference standard for analytical and metabolic research. It is intended for use as an internal standard or tracer in mass spectrometry and metabolic flux assays, supplied in small quantities suitable for laboratory workflows.
  • High isotopic enrichment and 99.7% chemical purity.
  • Suitable as an internal standard for GC/MS and LC/MS analyses.
  • Useful for metabolic tracing and flux experiments.
  • Available in small, precise quantities for laboratory use.
  • Stable crystalline form with defined molecular weight for reliable quantitation.
Catalog No. 50-002-13882 Supplier Medchemexpress LLC Supplier No. HY100582S25MG
Add to Cart
Add to Cart
Ribitol-3-13C is a 13C-labeled ribitol provided as a stable isotope reference standard for analytical and metabolic research. It is intended for use as an internal standard or tracer in mass spectrometry and metabolic flux assays, supplied in small quantities suitable for laboratory workflows.
  • High isotopic enrichment and 99.7% chemical purity.
  • Suitable as an internal standard for GC/MS and LC/MS analyses.
  • Useful for metabolic tracing and flux experiments.
  • Available in small, precise quantities for laboratory use.
  • Stable crystalline form with defined molecular weight for reliable quantitation.