Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ribitol-3-13C | 488-81-3 | 99.7% | 153.14 g/mol | C4 13C1 H12 O5 | 50 MG
SDP

Catalog No. 5000213661
Encompass_Preferred

Ribitol-3-13C is a 13C-labeled form of ribitol (adonitol) used as an isotopic tracer and internal standard for metabolic and biochemical analyses. Supplied in analytical pack sizes, the material is accompanied by a datasheet, certificate of analysis, and safety data sheet for lab use.

  • 3-13C isotopically labeled ribitol for tracer studies.
  • High purity: 99.7%.
  • Molecular weight ~153.14 g/mol, molecular formula C4 13C1 H12 O5.
  • Suitable for mass spectrometry and NMR applications.
  • Provided in small pack sizes for analytical use (e.g., 50 mg).
  • Comes with datasheet, certificate of analysis, and safety data sheet.

Catalog No. 50-002-13661 Supplier Medchemexpress LLC Supplier No. HY100582S250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ribitol-3-13C is a 13C-labeled form of ribitol (adonitol) used as an isotopic tracer and internal standard for metabolic and biochemical analyses. Supplied in analytical pack sizes, the material is accompanied by a datasheet, certificate of analysis, and safety data sheet for lab use.

  • 3-13C isotopically labeled ribitol for tracer studies.
  • High purity: 99.7%.
  • Molecular weight ~153.14 g/mol, molecular formula C4 13C1 H12 O5.
  • Suitable for mass spectrometry and NMR applications.
  • Provided in small pack sizes for analytical use (e.g., 50 mg).
  • Comes with datasheet, certificate of analysis, and safety data sheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.