Learn More
Medchemexpress LLC Ribitol-3-13C (Adonitol-3-13C) | 488-81-3 | 99.7% | 153.14 g·mol⁻1 | C5H12O5 (3-13C) | 10 MG
SDP
Catalog No. 5000213880
Encompass_Preferred
Ribitol-3-13C is ribitol labeled with a single 13C isotope at the 3-position, supplied as a high-purity stable-isotope reference standard for analytical and metabolic studies. It is a crystalline pentose alcohol commonly used as an internal standard in mass spectrometry and for isotope tracing in metabolic flux experiments.
  • Single 13C label at the 3-position for precise isotope tracing.
  • High purity (99.7%) for accurate quantitation and low background.
  • Suitable as an internal standard for LC-MS and GC-MS analyses.
  • Useful for metabolic flux analysis and pentose phosphate pathway studies.
  • Water-soluble crystalline material supplied in small mg quantities for analytical use.
Catalog No. 50-002-13880 Supplier Medchemexpress LLC Supplier No. HY100582S210MG
Ribitol-3-13C is ribitol labeled with a single 13C isotope at the 3-position, supplied as a high-purity stable-isotope reference standard for analytical and metabolic studies. It is a crystalline pentose alcohol commonly used as an internal standard in mass spectrometry and for isotope tracing in metabolic flux experiments.
  • Single 13C label at the 3-position for precise isotope tracing.
  • High purity (99.7%) for accurate quantitation and low background.
  • Suitable as an internal standard for LC-MS and GC-MS analyses.
  • Useful for metabolic flux analysis and pentose phosphate pathway studies.
  • Water-soluble crystalline material supplied in small mg quantities for analytical use.