Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ribitol-3-13C (adonitol-3-13C) | 488-81-3 | 99.7% | 153.14 g/mol | C4(13C)H12O5 | 1 MG
SDP

Catalog No. 5000213881
Encompass_Preferred

Ribitol-3-13C is a stable isotope-labelled ribitol supplied as a crystalline standard for use in analytical and metabolic studies. It is intended for use as a tracer or internal standard in NMR, GC-MS, and LC-MS workflows and is provided in a microgram-scale package suitable for analytical applications.

  • Contains a 13C label at the 3 position for isotope tracing.
  • High reported purity of 99.7% for analytical reliability.
  • Molecular weight approximately 153.14 g/mol for method calculations.
  • Provided as a crystalline solid for easy handling and storage.
  • Suitable as an internal standard for quantitative NMR, GC-MS, and LC-MS.
  • Supplied in a 1 mg quantity appropriate for small-scale analyses.

Catalog No. 50-002-13881 Supplier Medchemexpress LLC Supplier No. HY100582S21MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ribitol-3-13C is a stable isotope-labelled ribitol supplied as a crystalline standard for use in analytical and metabolic studies. It is intended for use as a tracer or internal standard in NMR, GC-MS, and LC-MS workflows and is provided in a microgram-scale package suitable for analytical applications.

  • Contains a 13C label at the 3 position for isotope tracing.
  • High reported purity of 99.7% for analytical reliability.
  • Molecular weight approximately 153.14 g/mol for method calculations.
  • Provided as a crystalline solid for easy handling and storage.
  • Suitable as an internal standard for quantitative NMR, GC-MS, and LC-MS.
  • Supplied in a 1 mg quantity appropriate for small-scale analyses.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.