Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ribitol | 488-81-3 | MFCD00064291 | ≥98.0% | 152.15 g/mol | C5H12O5 | 25 G
SDP

Catalog No. 5000211782
Encompass_Preferred

Ribitol is a crystalline pentose alcohol used as an analytical reagent and metabolic pathway standard. It is commonly employed in biochemical research to study the pentose phosphate pathway and to serve as an endogenous metabolite or analytical standard. The compound is solid at room temperature and is routinely prepared into stock solutions for laboratory use.

  • Crystalline pentose alcohol suitable for analytical use
  • Used in metabolic pathway and endogenous metabolite studies
  • Molecular formula C5H12O5; molecular weight 152.15 g/mol
  • Supplied as a solid for preparation of stock solutions
  • Storage guidance: powder stable at -20°C for years; in solvent store at -80°C for long-term

Catalog No. 50-002-11782 Supplier Medchemexpress LLC Supplier No. HY10058225G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ribitol is a crystalline pentose alcohol used as an analytical reagent and metabolic pathway standard. It is commonly employed in biochemical research to study the pentose phosphate pathway and to serve as an endogenous metabolite or analytical standard. The compound is solid at room temperature and is routinely prepared into stock solutions for laboratory use.

  • Crystalline pentose alcohol suitable for analytical use
  • Used in metabolic pathway and endogenous metabolite studies
  • Molecular formula C5H12O5; molecular weight 152.15 g/mol
  • Supplied as a solid for preparation of stock solutions
  • Storage guidance: powder stable at -20°C for years; in solvent store at -80°C for long-term

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.