Learn More
Medchemexpress LLC Ribitol | 488-81-3 | MFCD00064291 | 99.9% | 152.15 | C5H12O5 | 250 MG
SDP
Catalog No. 5000211575
Encompass_Preferred
Ribitol (adonitol) is a crystalline pentose alcohol supplied as a high-purity analytical standard for qualitative and quantitative laboratory analyses. It serves as a reference material for method development, calibration, and validation in chromatographic and mass spectrometric workflows.
  • High purity reference material for accurate quantitation.
  • Suitable for HPLC, GC, and MS method development.
  • Provides consistent calibration and quality control.
  • Available in multiple pack sizes for flexible use.
  • Solid crystalline form simplifies handling and storage.
Catalog No. 50-002-11575 Supplier Medchemexpress LLC Supplier No. HY100582R250MG
Ribitol (adonitol) is a crystalline pentose alcohol supplied as a high-purity analytical standard for qualitative and quantitative laboratory analyses. It serves as a reference material for method development, calibration, and validation in chromatographic and mass spectrometric workflows.
  • High purity reference material for accurate quantitation.
  • Suitable for HPLC, GC, and MS method development.
  • Provides consistent calibration and quality control.
  • Available in multiple pack sizes for flexible use.
  • Solid crystalline form simplifies handling and storage.