Learn More
VAZYME INTERNATIONAL LLC Vazyme International LLC Ribo-off rRNA Depletion Kit, Human/Mouse/Rat, 24 reactions
Catalog No. NC3979704
Encompass
This kit is designed for removing ribosomal RNA (rRNA) (including cytoplasmic 28S, 18S, 5.8S, 5S rRNA, and mitochondrial 16S, 12S rRNA) from total RNA of humans, mice, and rats, while retaining mRNA and other non-coding RNA. It is applicable to both intact and partially degraded RNA samples (e.g., FFPE RNA). The resulting ribosomal-depleted RNA can be used for the analysis of mRNA and non-coding RNA (such as lncRNA).
  • Wide species compatibility.
  • Complete retention of mRNA and lncRNA.
  • rRNA removal efficiency as high as 99.9%.
  • Removes cytoplasmic (28S, 18S, 5.8S, 5S rRNA) and mitochondrial (16S, 12S rRNA) ribosomal RNA.
  • Applicable to intact and partially degraded RNA samples (e.g., FFPE RNA).
  • Ribosomal-depleted RNA can be used for mRNA and non-coding RNA (e.g., lncRNA) analysis.
Catalog No. NC3979704 Supplier VAZYME INTERNATIONAL LLC Supplier No. N40601
This kit is designed for removing ribosomal RNA (rRNA) (including cytoplasmic 28S, 18S, 5.8S, 5S rRNA, and mitochondrial 16S, 12S rRNA) from total RNA of humans, mice, and rats, while retaining mRNA and other non-coding RNA. It is applicable to both intact and partially degraded RNA samples (e.g., FFPE RNA). The resulting ribosomal-depleted RNA can be used for the analysis of mRNA and non-coding RNA (such as lncRNA).
  • Wide species compatibility.
  • Complete retention of mRNA and lncRNA.
  • rRNA removal efficiency as high as 99.9%.
  • Removes cytoplasmic (28S, 18S, 5.8S, 5S rRNA) and mitochondrial (16S, 12S rRNA) ribosomal RNA.
  • Applicable to intact and partially degraded RNA samples (e.g., FFPE RNA).
  • Ribosomal-depleted RNA can be used for mRNA and non-coding RNA (e.g., lncRNA) analysis.