Learn More
Apexbio Technology LLC Ribosomal protein L3 peptide (202-222) amide 5mg
SDP
Catalog No. 5000063068
Encompass_Preferred
Ribosomal protein L3 peptide (202-222) amide is a synthetic peptide corresponding to residues 202 to 222 of ribosomal protein L3 with the sequence Met-Ser-His-Arg-Lys-Tyr-Glu-Ala-Pro-Arg-His-Gly-His-Leu-Gly-Phe-Leu-Pro-Arg-Lys-Arg-amide and a molecular formula of C114H182N42O25S (Mw 2573) Ribosomal protein L3 is an integral component of the large ribosomal subunit (60S) interacting with 23S rRNA to constitute the ribosome s peptidyl transferase center This particular peptide segment can bind to HIV-1 TAR RNA suggesting its role in studying TAR-mediated transcriptional activation and RNA-protein interactions in molecular virology research
Catalog No. 50-000-63068 Supplier Apexbio Technology LLC Supplier No. A10885
Ribosomal protein L3 peptide (202-222) amide is a synthetic peptide corresponding to residues 202 to 222 of ribosomal protein L3 with the sequence Met-Ser-His-Arg-Lys-Tyr-Glu-Ala-Pro-Arg-His-Gly-His-Leu-Gly-Phe-Leu-Pro-Arg-Lys-Arg-amide and a molecular formula of C114H182N42O25S (Mw 2573) Ribosomal protein L3 is an integral component of the large ribosomal subunit (60S) interacting with 23S rRNA to constitute the ribosome s peptidyl transferase center This particular peptide segment can bind to HIV-1 TAR RNA suggesting its role in studying TAR-mediated transcriptional activation and RNA-protein interactions in molecular virology research