Learn More
Medchemexpress LLC HY-15777 10mg Medchemexpress, Ribociclib CAS:1211441-98-3 Purity:>98%
SDP
Catalog No. 502029171
Encompass_Preferred
Medchemexpress, HY-15777 10mg Ribociclib CAS:1211441-98-3 Ribociclib (LEE01) is a highly specific CDK4/6 inhibitor with IC50 values of 10 nM and 39 nM, respectively, and is over 1,000-fold less potent against the cyclin B/CDK1 complex. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9171 Supplier Medchemexpress LLC Supplier No. HY1577710MG
Medchemexpress, HY-15777 10mg Ribociclib CAS:1211441-98-3 Ribociclib (LEE01) is a highly specific CDK4/6 inhibitor with IC50 values of 10 nM and 39 nM, respectively, and is over 1,000-fold less potent against the cyclin B/CDK1 complex. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.