Learn More
Selleck Chemical LLC Ribociclib
SDP
Catalog No. 5000004580
Encompass_Preferred
Ribociclib is an orally available and highly specific inhibitor of CDK4 and CDK6 with IC50 of 10 nM and 39 nM Phase 3
Catalog No. 50-000-04580 Supplier Selleck Chemical LLC Supplier No. S744025MG
Ribociclib is an orally available and highly specific inhibitor of CDK4 and CDK6 with IC50 of 10 nM and 39 nM Phase 3