Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ribociclib-d6 | 1328934-40-2 | 99.9% | 440.57 g/mol | C23H24D6N8O | 10 MG
SDP

Catalog No. 5000287902
Encompass_Preferred

Ribociclib-d6 is a deuterium-labeled isotopologue used as a stable isotope tracer and internal standard for quantitative bioanalytical methods such as NMR, GC-MS, and LC-MS.

  • Deuterium-labeled internal standard for NMR, GC-MS, and LC-MS quantitative analysis.
  • High chemical and isotopic purity (99.9%).
  • Molecular formula C23H24D6N8O; molecular weight 440.57 g/mol.
  • Solid, off-white to yellow appearance; provided as a 10 MG vial.
  • Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-87902 Supplier Medchemexpress LLC Supplier No. HY15777S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ribociclib-d6 is a deuterium-labeled isotopologue used as a stable isotope tracer and internal standard for quantitative bioanalytical methods such as NMR, GC-MS, and LC-MS.

  • Deuterium-labeled internal standard for NMR, GC-MS, and LC-MS quantitative analysis.
  • High chemical and isotopic purity (99.9%).
  • Molecular formula C23H24D6N8O; molecular weight 440.57 g/mol.
  • Solid, off-white to yellow appearance; provided as a 10 MG vial.
  • Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.