Learn More
Medchemexpress LLC HY-15777AS 5mg , Ribociclib-d6 (hydrochloride) CAS: Purity:>98%
SDP
Catalog No. 502260947
Encompass_Preferred
HY-15777AS 5mg Ribociclib-d6 (hydrochloride) CAS: Ribociclib D6 (LEE011 D6) hydrochloride is a deuterium labeled Ribociclib. Ribociclib is a highly specific CDK4/6 inhibitor with IC50 values of 10 nM and 39 nM, respectively, and is over 1,000-fold less potent against the cyclin B/CDK1 complex. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-0947 Supplier Medchemexpress LLC Supplier No. HY15777AS5MG
HY-15777AS 5mg Ribociclib-d6 (hydrochloride) CAS: Ribociclib D6 (LEE011 D6) hydrochloride is a deuterium labeled Ribociclib. Ribociclib is a highly specific CDK4/6 inhibitor with IC50 values of 10 nM and 39 nM, respectively, and is over 1,000-fold less potent against the cyclin B/CDK1 complex. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.